missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl) Disulfide 98.0+%, TCI America™

Catalog No. B426325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B426325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B426325G Supplier TCI America Supplier No. B426325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1892-29-1
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00002906
InChI Key KYNFOMQIXZUKRK-UHFFFAOYSA-N
Synonym 2-hydroxyethyl disulfide, 2,2'-dithiodiethanol, bis 2-hydroxyethyl disulfide, ethanol, 2,2'-dithiobis, hydroxyethyl disulfide, diethanol disulfide, 2,2'-dithiobisethanol, 2-hydroxyethanedisulfide, usaf th-9, 2,2'-disulfanediyldiethanol
PubChem CID 15906
ChEBI CHEBI:43136
IUPAC Name 2-[(2-hydroxyethyl)disulfanyl]ethan-1-ol
SMILES OCCSSCCO

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 g
UN Number 2810
Formula Weight 154.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-hydroxyethyl) Disulfide
RTECSNumber KK7525000
TSCA Yes