missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl)amine, 99%

Catalog No. AAL13894AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13894AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL13894AP Supplier Thermo Scientific Chemicals Supplier No. L13894AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-20-7 | C16H35N | 241.463 g/mol

Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl)amine (BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules

Solubility
Soluble in water 0.045 g/L @ 20˚C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 106-20-7
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
MDL Number MFCD00009489
InChI Key SAIKULLUBZKPDA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619
PubChem CID 7791
IUPAC Name 2-ethyl-N-(2-ethylhexyl)hexan-1-amine
SMILES CCCCC(CC)CNCC(CC)CCCC

Specifications

Melting Point <-60°C
Density 0.805
Boiling Point 281°C
Flash Point 121°C (250°F)
Refractive Index 1.442
Quantity 500 mL
UN Number UN2922
Beilstein 1748342
Solubility Information Soluble in water 0.045g/L @ 20?C.
Formula Weight 241.46
Percent Purity 99%
Chemical Name or Material Bis(2-ethylhexyl)amine
Show More Show Less
Hazard Category H302-H311+H331-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-372-4
RTECSNumber IH6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only