missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) adipate, 99%, MilliporeSigma™ Supelco™

Catalog No. 111012338
Change view
Click to view available options
Quantity:
5 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012338 5 mL
111012337 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012338 Supplier Merck Emd Millipore Supplier No. 5251975ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00009496
Synonym Adipic acid di(2-ethylhexyl) ester; DOA

Specifications

Melting Point <-70°C
CAS 103-23-1
Density 0.925 g/mL (at 20°C)
Boiling Point 175°C (2 mmHg)
Flash Point 196°C
Packaging Glass Bottle
Linear Formula [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 5 mL
Formula Weight 370.57
Percent Purity 99%