missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-carboxyethyl) Isocyanurate 98.0+%, TCI America™

Catalog No. I0584500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0584500G 500 g
I058425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0584500G Supplier TCI America Supplier No. I0584500G
Only null left

Chemical Identifiers

CAS 2904-40-7
Molecular Formula C9H11N3O7
Molecular Weight (g/mol) 273.201
MDL Number MFCD04038422
InChI Key JCUMQTAAEUDUPK-UHFFFAOYSA-N
Synonym Isocyanuric Acid Bis(2-carboxyethyl) Ester
PubChem CID 223763
IUPAC Name 3-[3-(2-carboxyethyl)-2,4,6-trioxo-1,3,5-triazinan-1-yl]propanoic acid
SMILES C(CN1C(=O)NC(=O)N(C1=O)CCC(=O)O)C(=O)O

Specifications

Melting Point 289°C
Color White
Quantity 500 g
Formula Weight 273.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-carboxyethyl) Isocyanurate
EINECSNumber (5)-1063
TSCA No