missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-butoxyethyl) Ether, 98.5%, Spectrum™ Chemical

Catalog No. 18609078
Change view
Click to view available options
Quantity:
100 mL
500 mL
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609078 500 mL Amber Glass Bottle
18609075 100 mL Amber Glass Bottle
18609076 20 L Amber Glass Bottle
18609077 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609078 Supplier Spectrum Chemical Mfg Cor Supplier No. B1631500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 112-73-2

Specifications

Acidity 100 ppm
CAS Max % 1
Boiling Point 256°C (492.8°F)
Packaging Amber Glass Bottle
Quantity 500 mL
Molecular Weight (g/mol) 218.38
Percent Purity 0.985
Grade Reagent
Physical Form Liquid
Chemical Name or Material Bis(2-butoxyethyl) Ether

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.