missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)palladium(II) 98.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 1 g |
| Formula Weight | 304.64 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(2,4-pentanedionato)palladium(II) |
Sécurité et manipulation
| EINECSNumber | (2)-2181 |
| TSCA | Yes |