missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)nickel(II) Hydrate 98.0+%, TCI America™

Catalog No. N0096500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0096500G 500 g
N009625G 25 g
N0096100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0096500G Supplier TCI America Supplier No. N0096500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3264-82-2
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
PubChem CID 53384569
IUPAC Name nickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Color Blue
Quantity 500 g
Formula Weight 256.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)nickel(II) Hydrate
EINECSNumber (2)-2181
RTECSNumber SA2100000
TSCA Yes