missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)magnésium(II) 98,0+%, TCI America™

Catalog No. M00015G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
M00015G 5 g
M000125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00015G Supplier TCI America Supplier No. M00015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14024-56-7
Formule moléculaire C10H14MgO4
Poids moléculaire (g/mol) 222.523
Numéro MDL MFCD00013497
Clé InChI AKTIAGQCYPCKFX-SYWGCQIGSA-L
Synonyme 4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one
PubChem CID 25113476
Nom de l’IUPAC magnésium; (E)-4-oxopent-2-en-2-olate
SOURIRES CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]

Specifications

Nom ou substance chimique Bis(2,4-pentanedionato)magnesium(II)
Point de fusion 276 °C
Couleur Blanc
Quantité 5 g
Poids de formule 222.52
Pourcentage de pureté ≥98,0% (T)
Forme physique Poudre cristalline
EINECSNumber (2)-2181
TSCA Non