missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-dinitrophenyl) Oxalate 98.0+%, TCI America™

Catalog No. A53035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53035G 5 g
A53031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53035G Supplier TCI America Supplier No. A53035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Chemiluminescence reagent for the determination of fluorescent compounds by HPLC and FIA]

Chemical Identifiers

CAS 16536-30-4
Molecular Formula C14H6N4O12
Molecular Weight (g/mol) 422.218
MDL Number MFCD00010712
InChI Key CBZOGAWUNMFXFQ-UHFFFAOYSA-N
Synonym DNPO, Oxalic Acid Bis(2,4-dinitrophenyl) Ester
PubChem CID 3080704
IUPAC Name bis(2,4-dinitrophenyl) oxalate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])OC(=O)C(=O)OC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 422.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-dinitrophenyl) Oxalate [Chemiluminescence reagent for the determination of fluorescent compo