missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4,6-trichlorophenyl)oxalate, 98%

Catalog No. AC402660050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402660050 Supplier Thermo Scientific Chemicals Supplier No. 402660050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1165-91-9 | C14H4Cl6O4 | 448.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1165-91-9
Molecular Formula C14H4Cl6O4
Molecular Weight (g/mol) 448.89
MDL Number MFCD00043061
InChI Key GEVPIWPYWJZSPR-UHFFFAOYSA-N
Synonym bis 2,4,6-trichlorophenyl oxalate, bis 2,4,6-trichlorophenyl ethanedioate, oxalic acid bis 2,4,6-trichlorophenyl ester, 2,4,6-trichlorophenyl oxalate, ethanedioic acid, bis 2,4,6-trichlorophenyl ester, bis-tcpo, acmc-20a7jt, gevpiwpywjzspr-uhfffaoysa
PubChem CID 160567
IUPAC Name bis(2,4,6-trichlorophenyl) oxalate
SMILES C1=C(C=C(C(=C1Cl)OC(=O)C(=O)OC2=C(C=C(C=C2Cl)Cl)Cl)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 188°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 448.89
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2, 4, 6-trichlorophenyl)oxalate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only