missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)

Catalog No. AA4135909
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4135909 10 g
AA4135902 0.5 g
AA4135904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4135909 Supplier Thermo Scientific Chemicals Supplier No. 04135909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Precursor for CVD process | CAS: 13986-53-3 | C22H38CoO4 | 425.48 g/mol

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13986-53-3
Molecular Formula C22H38CoO4
Molecular Weight (g/mol) 425.48
MDL Number MFCD00233616
InChI Key KLJJOSZRALJWDS-UHFFFAOYSA-N
Synonym bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
PubChem CID 57369563
IUPAC Name cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES [Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 143°C
Quantity 10 g
Solubility Information Soluble in hydrocarbons,alcohols and ethers. Slightly soluble in water.
Formula Weight 425.46
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)
Hazard Category H317-H341
Hazard Statement GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only