missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Sebacate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 222°C |
| Quantity | 5 g |
| Formula Weight | 508.79 |
| Percent Purity | ≥95.0% (GC) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Sebacate |
Safety and Handling
| EINECSNumber | (5)-5501 |
| TSCA | Yes |