Recombinant Protein;25UG;Biotinylated Human CA125 (13810-14451), Fc,Avitag (CA5-H82F8) is expressed from human 293 cells (HEK293). It contains AA Pro 13810 - Pro 14451 (Accession # Q8WXI7-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 100.0 kDa. The protein migrates as 120-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.