Learn More
ACROBiosystems Inc Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag™&Flag Tag (MALS verified)
Numéro de catalogue. 502054879
Encompass
Recombinant Protein;25UG;Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag (CDG-C8219) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G)).;Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 17.7 kDa (CD3E) and 15.4 kDa (CD3G). Subunit CD3E carries a polyhistidine tag at the C-terminus, followed by an Avi tag and subunit CD3G is fused with a flag tag at the C-terminus. As a result of glycosylation, the protein migrates as 19-20 kDa and 22-24 kDa under reducing (R) condition, and 35-40 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Bispecific T cell Engager (CD3 X BCMA) (Cat. No. CDA-H52Hb) at 1g/mL (100L/well) can bind Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag (Cat. No.CDG-C8219) with a linear range of 0.98-15.6 ng
Numéro de catalogue. 502054879 Fournisseur ACROBiosystems Inc Code fournisseur CDGC821925UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;25UG;Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag (CDG-C8219) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (CD3E) & Gln 23 - Thr 113 (CD3G) (Accession # Q95LI5-1 (CD3E) & Q95LI7-1 (CD3G)).;Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 17.7 kDa (CD3E) and 15.4 kDa (CD3G). Subunit CD3E carries a polyhistidine tag at the C-terminus, followed by an Avi tag and subunit CD3G is fused with a flag tag at the C-terminus. As a result of glycosylation, the protein migrates as 19-20 kDa and 22-24 kDa under reducing (R) condition, and 35-40 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Bispecific T cell Engager (CD3 X BCMA) (Cat. No. CDA-H52Hb) at 1g/mL (100L/well) can bind Biotinylated Cynomolgus CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag (Cat. No.CDG-C8219) with a linear range of 0.98-15.6 ng