missing translation for 'onlineSavingsMsg'
Learn More

Biotin, Powder, USP, 97.5-102%, Spectrum™ Chemical

Catalog No. 18600995
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600995 1 kg Amber Glass Bottle
18600989 500 mg Amber Glass Bottle
18600990 1 g Amber Glass Bottle
18600991 5 g Amber Glass Bottle
18600992 25 g Amber Glass Bottle
18600993 100 g Amber Glass Bottle
18600994 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18600995 Supplier Spectrum Chemical Mfg Cor Supplier No. B11031KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

B1103, 58-85-5, C10H16N2O3S

Spectrum™ Chemical Biotin, Powder, USP is used for preventing and treating biotin deficiency. Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Specific Rotation +89° to +93°
Percent Purity 97.5 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Biotin Powder