missing translation for 'onlineSavingsMsg'
Learn More

Biotin, Powder, FCC, 97.5-100.5%, Spectrum™ Chemical

Catalog No. 18600983
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600983 500 mg Amber Glass Bottle
18600984 1 g Amber Glass Bottle
18600985 5 g Amber Glass Bottle
18600986 25 g Amber Glass Bottle
18600987 100 g Amber Glass Bottle
18600988 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600983 Supplier Spectrum Chemical Mfg Cor Supplier No. BI115500MG
Get Up to $500 in Chemicals with Purchase
View all promotions

BI115, 58-85-5, C10H16N2O3S

Spectrum™ Chemical Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

CAS Max % 100%
Melting Range 229-232°C
Packaging Amber Glass Bottle
Quantity 500 mg
Optical Rotation +89° to +93°
Percent Purity 97.5 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Biotin Powder