missing translation for 'onlineSavingsMsg'
Learn More

Biotine-PEG3-Amine 95,0+%, TCI America™

Catalog No. B317225MG
Change view
Click to view available options
Quantité:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B317225MG 25 mg
B3172100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B317225MG Supplier TCI America Supplier No. B317225MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 359860-27-8
Formule moléculaire C18H34N4O5S
Poids moléculaire (g/mol) 418.553
Numéro MDL MFCD09952640
Clé InChI KIJSBKNJFAUJFV-ZOBUZTSGSA-N
Synonyme N-Biotinyl-3,6,9-trioxaundecane-1,11-diamine, N-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethyl]biotinamide
PubChem CID 50896259
Nom de l’IUPAC 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-(2-amino-ethoxy)ethoxy]éthyl]pentanamide
SOURIRES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN)NC(=O)N2

Specifications

Nom ou substance chimique Biotin-PEG3-Amine
Couleur Blanc
Quantité 25 mg
Poids de formule 418.55
Pourcentage de pureté ≥95,0% (HPLC)
Forme physique Poudre cristalline
TSCA Non
Recommended Storage Congélateur