missing translation for 'onlineSavingsMsg'
Learn More

Biotin-PEG2-Amine 98.0+%, TCI America™

Catalog No. B317125MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B317125MG 25 mg
B3171100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B317125MG Supplier TCI America Supplier No. B317125MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138529-46-1
Molecular Formula C16H30N4O4S
Molecular Weight (g/mol) 374.50
MDL Number MFCD09264745
InChI Key LWISPDYGRSGXME-UHFFFAOYNA-N
Synonym N-Biotinyl-3,6-dioxaoctane-1,8-diamine, N-[2-[2-(2-Aminoethoxy)ethoxy]ethyl]biotinamide
PubChem CID 11199678
IUPAC Name N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
SMILES NCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Color White
Quantity 25 mg
Formula Weight 374.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin-PEG2-Amine
TSCA No
Recommended Storage Freezer