Learn More
ACROBiosystems Inc Recombinant Protein;Biotinylated Human CD47 Protein, Avi Tag (Avitag™);HEK293;200UG;CD7,CD47,MER6,IAP,OA3
Numéro de catalogue. 501253822
Encompass
Recombinant Protein;Biotinylated Human CD47 Protein, Avi Tag (Avitag™);200UG;Biotinylated Human CD47, His Tag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 16.9 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD47 MAb, Human IgG4 at 2 g/mL (100 L/well) can bind Biotinylated Human CD47, His Tag (Cat. No. CD7-H82E9) with a linear range of 0.07-0.84 ng/mL (QC tested).;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2 g/mL (100 L/well) can bind Biotinylated Human CD47, His Tag (Cat. No. CD7-H82E9) with a linear range of 0.8-13.1 ng/mL (Routinely tested).
Numéro de catalogue. 501253822 Fournisseur ACROBiosystems Inc Code fournisseur CD7H82E9200UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Biotinylated Human CD47 Protein, Avi Tag (Avitag™);200UG;Biotinylated Human CD47, His Tag (CD7-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 16.9 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD47 MAb, Human IgG4 at 2 g/mL (100 L/well) can bind Biotinylated Human CD47, His Tag (Cat. No. CD7-H82E9) with a linear range of 0.07-0.84 ng/mL (QC tested).;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2 g/mL (100 L/well) can bind Biotinylated Human CD47, His Tag (Cat. No. CD7-H82E9) with a linear range of 0.8-13.1 ng/mL (Routinely tested).