Learn More
Biosynergy HemoTemp™ Reversible Blood Temperature Indicator
Catalog No. 22246193
Encompass
Catalog No. 22246193 Supplier Biosynergy Supplier No. 2000B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Shows the internal temperature of blood bags under ambient conditions
  • Indicator has a temperature range of 1° to 12°C and gives an instant temperature readout at a specific point in time
  • Does not indicate if the blood has exceeded the maximum permissible temperature and then recooled since the indicator is reversible
  • Should not be the sole method for determining the clinical acceptability of blood
Specifications
For Use With (Application) Utilize liquid crystal technology and offer reversible temperature readouts
Product Type Reversible Blood Bag Temperature Indicator