Learn More
Biosynergy HemoTemp™ II Reversible/Irreversible Blood Temperature Indicator
Catalog No. 22246194
Encompass
Catalog No. 22246194 Supplier Biosynergy Supplier No. 3000B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reversible and irreversible indicator
  • Reversible indicator acts as a blood bag thermometer calibrated to display the internal temperature of the bag under ambient conditions
  • The irreversible portion signals that the internal blood bag temperature has reached or exceeded 10°C; the signal continues even if the blood is recooled
  • Should not be the sole method for evaluating the clinical acceptability of blood
Specifications
For Use With (Application) Time and temperature history of the blood unit
Product Type Blood bag temperature indicator