missing translation for 'onlineSavingsMsg'
Learn More

Innovatek Medical Inc. BioStrip hCG Urine Pregnancy Test Strips

Catalog No. 361070780
Change view
Click to view available options
Packaging Type:
Desiccated Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
361070780 Desiccated Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361070780 Supplier Innovatek Medical Inc. Supplier No. 422050/DUP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect hCG in urine with a visual, qualitative test for professional use. Sensitivity: 20 IU/L; 50 strips per kit.

  • Qualitative hCG detection in urine for early pregnancy detection
  • Dip for at least 10 seconds without immersing past the MAX line
  • Lay flat after dipping; allow the full 5 minutes to confirm a negative result
  • Do not read results after 10 minutes
  • Built-in procedural control band
  • Store at 2-30°C in the original desiccated container with the lid tightly closed; do not freeze
  • 50 strips per kit; external hCG controls available separately

Specifications

Content And Storage Store At 2°C to 30°C In The Original Tightly Closed Desiccated Container. Do Not Freeze. Protect From Heat, Moisture, and Direct Sunlight.
CE Marker No
Clia Complexity Not Applicable
Detectable Analytes hCG
Format Test Strip
Sample Type Urine
Sensitivity 20 IU/L hCG
Type Urine Pregnancy Test Strip
Disposable Single-use
For Use With (Application) Visual, qualitative detection of hCG in urine for early pregnancy detection
Includes 50 test strips and instructions for use
Packaging Type Desiccated Container
Quantity 50 Strips/kit
Test Time 5 Minutes to Confirm A Negative Result
Form Solid
Operating Temperature Range 20°C to 30°C
Show More Show Less