missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIOshell™ IgG 1000 Å Diphenyl, 2.7 μm HPLC Guard Cartridge, Sold by MilliporeSigma™ Supelco™
Liquid chromatography column based on a new wide-pore
$1108.49 - $1108.49
Specifications
| Quantity | 3/Pk. |
|---|---|
| pH | 2 to 9 |
| Particle Size | 2.7 μm |
| Length (Metric) | 0.5 cm |
| Pore Size | 1000 Å pore size |
Description
- High-speed, high-performance liquid chromatography column based on a new wide-pore (1000 Å) Fused-Core™ particle design.
- The particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
- This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm.
- The densely bonded, extensively endcapped diphenylmethyl bonded phase of BIOshell™IgG 1000 Å Diphenyl provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.
Specifications
| 3/Pk. | |
| 2.7 μm | |
| 1000 Å pore size | |
| HPLC: Suitable, L11 Per USP | |
| Endcapped |
| 2 to 9 | |
| 0.5 cm | |
| 22 m2/g | |
| Bonding phase, Diphenyl (diphenylmethyl) | |
| Fused-Core™ particle platform |
BIOshell is a trademark of Sigma-Aldrich Co. LLC, Fused-Core is a registered trademark of Advanced Materials Technology, Inc.