missing translation for 'onlineSavingsMsg'
Learn More

BIOshell™ IgG 1000 Å Diphenyl, 2.7 μm HPLC Guard Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022284
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111022284 2.1 mm
111022285 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022284 Supplier Merck Emd Millipore Supplier No. 577431U

Liquid chromatography column based on a new wide-pore

  • High-speed, high-performance liquid chromatography column based on a new wide-pore (1000 Å) Fused-Core™ particle design.
  • The particle provides a thin porous shell of high-purity silica surrounding a solid silica core.
  • This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm.
  • The densely bonded, extensively endcapped diphenylmethyl bonded phase of BIOshell™IgG 1000 Å Diphenyl provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.

Specifications

Quantity 3/Pk.
pH 2 to 9
Particle Size 2.7 μm
Length (Metric) 0.5 cm
Pore Size 1000 Å pore size
Surface Area 22 m2/g
Diameter (Metric) Inner 2.1 mm
For Use With (Equipment) HPLC: Suitable, L11 Per USP
Stationary Phase Bonding phase, Diphenyl (diphenylmethyl)
Endcapped Endcapped
Matrix Fused-Core™ particle platform
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC, Fused-Core is a registered trademark of Advanced Materials Technology, Inc.