Learn More
Thermo Scientific™ BioProcess Container (BPC), 50 L, for HyPerforma™ Single-Use Mixer DS 300, Bottom drain, irradiated
Catalog No. SH3094601
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
SH3094601 Each
1 options
Catalog No. SH3094601 Supplier Thermo Scientific™ Supplier No. SH30946.01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 50 L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300.
The 50 L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.
Key features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:
  • All standard BPCs and liners are designed to fit 'out-of-the-box'
  • Available in volume options of 50, 100, and 200 L
  • Applicable to open- and closed-top mixing applications
  • Powder-liquid and liquid-liquid mixing
  • World class CX5-14 and CX3-9 film
  • Adaptable to multiple upstream process applications in media and buffer prep
Required accessories:
  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma SIngle-Use Mixer DS 300 drums
Recommended for:
  • Powder-liquid or liquid-liquid mixing applications in single use
  • High-throughput mixing and storage
  • Additional configurations are available to suit your application requirements.
Accessory
  • Reusable Bearing Hub (Cat No. SV5017777)
Specifications
Capacity (Metric) 50 L
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity Each
Product Line HyPerforma