A perfusable channel -Slide with 3x7 wells for long-term spheroid cultivation and high-end microscopy. Ideal for the long-term cultivation and perfusion of 3D spheroids or organoids on the Bioinert surface. Allowing for flexible preparation for perfusion, spheroids can be transferred or created directly in the wells. Imaging Chamber with excellent optical quality for high-resolution microscopy. Pcs./Box: 15 (individually packed)Surface Modification: Bioinert: #1.5 polymer coverslip, surface passivation with Bioinert, sterilized