missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Biohazard Disposal Bags
GREENER_CHOICE

Catalog No. 03448274
Change view
Click to view available options
Product Type:
Bag Holder
Benchtop Biohazard Disposal Bag
Capacity (Metric):
2 L
9 L
19 L
40 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Capacity (Metric)
03448274 Benchtop Biohazard Disposal Bag 40 L
03448271 Benchtop Biohazard Disposal Bag 2 L
03448272 Benchtop Biohazard Disposal Bag 9 L
03448273 Benchtop Biohazard Disposal Bag 19 L
03448270 Bag Holder 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03448274 Supplier Heathrow Scientific™ Supplier No. HS10323

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag

Specifications

Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With (Application) Biohazard Disposal
For Use With (Equipment) Autoclave
Material Polypropylene
Sterility Non-sterile
No. per Case 500 Pk.
Outer Packaging Box
Thickness (English) 63 mil
Dimensions (L x W) 25 x 33 in.
Dimensions (L x W x H) 25 x 33 x 0.06 in.
Length (English) 25 in.
Width (English) 33 in.
Height (English) 0.06 in.
Height (Metric) 0.15 cm
Weight (English) 0.08 lb.
Weight (Metric) 0.04 kg
Product Type Benchtop Biohazard Disposal Bag
Capacity (English) 10.6 gal.
Capacity (Metric) 40 L
Length (Metric) 63.5 cm
Width (Metric) 83.8 cm
Thickness (Metric) 1.6 mm
Show More Show Less