missing translation for 'onlineSavingsMsg'
Learn More

Biochanin A, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014310
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014310 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014310 Supplier Merck Emd Millipore Supplier No. 9214250MG
Only null left
Food and Beverages

Specifications

CAS 491-80-5
Linear Formula C16H12O5
MDL Number MFCD00006839
Melting Point 210°C to 213°C
Synonym 4 ′-Methylgenistein; 5,7-Dihydroxy-4 ′-methoxyisoflavone; Genistein 4 ′-methyl ether; Olmelin
Type Analytical Standard
Quantity 50 mg
Formula Weight 284.26
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard