Learn More
Simport™ Scientific Bioblock™ Polypropylene Deep Well Plates, U-bottom
All BioBlock™ Deep Well Plates are ideal for high volume, high throughput applications. These high-performance plates are chemical-resistant and are designed for automated robotic handling.
$210.98 - $643.98
Specifications
Lid Without Lid
Autoclavable Autoclavable
Binding Type Regular
Product Line Microtiter
Disposable Single-use
View More Specs
Products 17
Catalog Number Mfr. No. Color Dimensions (L x W x H) No. of Wells Volume (Metric) Well Price Quantity  
Catalog Number Mfr. No. Color Dimensions (L x W x H) No. of Wells Volume (Metric) Well Price Quantity  
01670692
View Documents
Simport™ Scientific
T11018
Clear 127.8 x 85.5 x 43.6 mm 48 3.5 mL
Case of 50 Each for $450.77
Only null left
01670680
View Documents
Simport™ Scientific
T1101
Clear 127.8 x 85.5 x 14.8 mm 96 400 μL
Case of 100 Each for $482.96
Only null left
01670703
View Documents
Simport™ Scientific
T11032
Clear 127.5 x 85.4 x 24.4 mm 96 1 mL
Case of 100 Each for $643.98
Only null left
01670683
View Documents
Simport™ Scientific
T1104
Clear 127.8 x 85.5 x 33 mm 96 1 mL
Case of 50 Each for $322.00
Only null left
361045528
View Documents
Simport™ Scientific
T1105
Clear 127.8 x 85.5 x 41.6 mm 96 1.2 mL
Case of 24 Each for $266.50
Only null left
01670686
View Documents
Simport™ Scientific
T1105B
Clear, Blue 127.8 x 85.5 x 41.6 mm 96 1.2 mL
Case of 24 Each for $210.98
Only null left
01670688
View Documents
Simport™ Scientific
T1105P
Clear, Red 127.8 x 85.5 x 41.6 mm 96 1.2 mL
Case of 24 Each for $210.98
Only null left
01670689
View Documents
Simport™ Scientific
T1105Y
Clear, Yellow 127.8 x 85.5 x 41.6 mm 96 1.2 mL
Case of 24 Each for $210.98
Only null left
01670691
View Documents
Simport™ Scientific
T1108
Clear 127.5 x 85.4 x 44.3 mm 96 2 mL
Case of 50 Each for $488.05
Only null left
01670695
View Documents
Simport™ Scientific
T11010B
Clear, Blue 127.8 x 85.5 x 41.6 mm 96 2.1 mL
Case of 24 Each for $275.75
Only null left
01670696
View Documents
Simport™ Scientific
T11010G
Clear, Green 127.8 x 85.5 x 41.6 mm 96 2.1 mL
Case of 24 Each for $275.75
Only null left
01670697
View Documents
Simport™ Scientific
T11010P
Clear, Red 127.8 x 85.5 x 41.6 mm 96 2.1 mL
Case of 24 Each for $275.75
Only null left
01670698
View Documents
Simport™ Scientific
T11010Y
Clear, Yellow 127.8 x 85.5 x 41.6 mm 96 2.1 mL
Case of 24 Each for $275.75
Only null left
01670708
View Documents
Simport™ Scientific
T11045
Clear 127.5 x 85.4 x 41.6 mm 96 2.2 mL
Case of 96 Each for $618.19
Only null left
01670706
View Documents
Simport™ Scientific
T11040
Clear 127 x 85 x 42.5 mm 96 2.2 mL
Case of 50 Each for $322.00
Only null left
01670701
View Documents
Simport™ Scientific
T11029
Clear 127.8 x 85.5 x 44.2 mm 48 4.6 mL
Case of 96 Each for $618.19
Only null left
01670704
View Documents
Simport™ Scientific
T11034
Clear 127.5 x 85.4 x 31.6 mm 96 1.6 mL
Case of 50 Each for $322.00
Only null left
Description
Description
  • Footprint conforms to SBS/ANSI standards
  • High chemical stability
  • Free of RNase, DNase, pyrogens, and DNA
  • Maximum sustainable centrifuge force up to 4000 g
  • Height difference from center to angle is less than 0.4 mm
  • Models are offered with 24, 48, 96, and 384 round or square-shaped wells
  • Clear wells are available with your choice of U-shaped or V-shaped bottoms
  • Available in a variety of working volumes; from 400 μL to 3.5 mL
Specifications
Specifications
Without Lid
Regular
Single-use
Polypropylene
Round Bottom (U-well)
Yes
Autoclavable
Microtiter
Robotics
Non-sterile
Yes
Certified Quality
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'