missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BIO

Catalog No. 319410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
319410 10 mg
319450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 319410 Supplier Tocris Bioscience™ Supplier No. 3194/10

Potent GSK-3 inhibitor; also inhibits cdks

Potent ATP-competitive glycogen synthase kinase-3 (GSK-3) inhibitor (IC50 = 5 nM). Also inhibits cyclin-dependent kinases (cdks; IC50 values are <100 nM for cdk2 and 5 and sub micromolar for cdk1, cdk4 and cdk9). Maintains self-renewal and pluripotency in embryonic stem cells via activation of the Wnt signaling pathway in vitro; also promotes proliferation and dedifferentiation in cardiomyocytes.

Negative Control also available.

Chemical Identifiers

CAS 667463-62-9
Molecular Formula C16H10BrN3O2

Specifications

Quantity 10 mg
Formula Weight 356.17
Purity >98%
Molecular Weight (g/mol) 356.179
Chemical Name or Material BIO
Recommended Storage Store at +4°C