accessibility menu, dialog, popup

UserName

CAS RN 667463-62-9

IUPAC Name: 6'-bromo-3-(hydroxyamino)-1',2'-dihydro-[2,3'-biindolyliden]-2'-one
Synonyms: 6'-bromo-3-(hydroxyamino)-1'H-[2,3'-biindolyliden]-2'-one
Molecular Weight (g/mol): 356.18
Molecular Formula: C16H10BrN3O2
InChi Key: WNWSUJQVZJJGLF-UHFFFAOYSA-N
SMILES: ONC1=C2C=CC=CC2=NC1=C1C(=O)NC2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)