missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader
GREENER_CHOICE

The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.

Supplier:  Agilent BioTek SLXFSN

 Afficher plus de versions de ce produit

Numéro de catalogue. BTSLXF

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability.

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately).

Certifications

CE and TUV marked. RoHS compliant.

Warranty

12 Months

Spécifications

Identifiants chimiques

200 to 900 nm
CE and TUV marked. RoHS compliant.
38.1 cm
24 sec. (96 wells), 76 sec. (384 wells)
Monochromator Bandpass
50/60 Hz
Multi-mode Microplate Reader
0.001 OD
2
1 year
27 lb.
15 in.
Photodiode, PMT
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
≤5 nm
15 in.
Linear, Orbital, Double Orbital
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
<1% from 0 to 2.5 OD
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
English
100/240 V
12.3 kg
38.1 cm
Xenon Flash

Spécifications

200 to 900 nm
CE and TUV marked. RoHS compliant.
38.1 cm
24 sec. (96 wells), 76 sec. (384 wells)
Monochromator Bandpass
12 in.
30.5 cm
Reader Only
SLXF
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
English
100/240 V
12.3 kg
38.1 cm
Gen5 Data Analysis Software
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
≤5 nm
15 in.
Linear, Orbital, Double Orbital
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
50/60 Hz
<1% from 0 to 2.5 OD
Fluorescence, Luminescence
Multi-mode Microplate Reader
0.001 OD
2
1 year
27 lb.
15 in.
Photodiode, PMT
Xenon Flash
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions