missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Fast-Trac Activity Wheel, Certified

Catalog No. 360993902
Encompass
Change view
Click to view available options
Color:
Amber
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
360993902 Blue
360993901 Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 360993902 Supplier Bio-Serv™ Supplier No. K3251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specialized activity wheel for use with the Mouse Igloo shelter (Mfr. Nos. K3328, K3570, and K3327)

  • Supports natural and instinctive running behavior
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)

Specifications

Autoclavable Autoclavable
Product Type Fast-Trac Wheel Activity
Color Blue
Material Polycarbonate
For Use With (Application) Supports natural and instinctive running behavior
For Use With (Equipment) Mouse igloo