Learn More
Research Products Corporation Bio-Safe NA Complete Counting Cocktail, 4 Liters
Numéro de catalogue. 50550146
Encompass
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers such as TS-2. Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.
Numéro de catalogue. 50550146 Fournisseur Research Products Corporation Code fournisseur RPI 111198
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers such as TS-2. Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.