Learn More
Innovatek BinaxNow Rapid Test Kits
Catalog No. 361045559
Change view
Click to view available options
Quantity:
22 Tests/Kit
Disease:
Influenza A and B
Catalog No. Quantity Disease
361045559 22 Tests/Kit Influenza A and B
1 options
Catalog No. 361045559 Supplier Innovatek Supplier No. 4010WL416000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Innovatek BinaxNow Rapid Test Kits provide results within 15 minutes
  • Influenza A & B Test Kit
    • Rapid detection of Influenza A and B Nucleoprotein Antigens
    • Used for nasopharyngeal (NP) swab, nasal swab samples, and nasal wash/aspirate specimens
    • Results in 15 minutes
  • Malaria Test Kit
    • Rapid detection of Plasmodium antigens circulating in human venous and capillary EDTA whole blood
    • Differential diagnosis of Plasmodium falciparum (P.f.) infections from other less virulent malarial infections
    • Results in 15 minutes
Specifications
Quantity 22 Tests/Kit
Disease Influenza A and B
Reaction Speed 15 min.