missing translation for 'onlineSavingsMsg'
Learn More

Abbott™ BinaxNOW™ Legionella Urinary Antigen Test Control Swab

Catalog No. BNX852010
Encompass
Change view
Click to view available options
Quantity:
Control Swab Pack (5 positive/5 negative)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BNX852010 Control Swab Pack (5 positive/5 negative)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BNX852010 Supplier Abbott™ Supplier No. 852010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the detection of Legionella pneumophila serogroup 1 antigen in urine specimens.

Order Info

Use with BinaxNOW™ Legionella Urinary Antigen Test Kits, Cat. No. BNX852000

Specifications

Sample Type Urine
For Use With (Application) Qualitatively detect Legionella pneumophila serogroup 1 in Urine using BinaxNow™ Legionella Urinary Antigen Test Kit
Quantity Control Swab Pack (5 positive/5 negative)