missing translation for 'onlineSavingsMsg'
Learn More

Bimatoprost, Thermo Scientific™

Catalog No. AC468881000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468881000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468881000 Supplier Thermo Scientific Chemicals Supplier No. 468881000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 155206-00-1
Molecular Formula C25H37NO4
Molecular Weight (g/mol) 415.57
MDL Number MFCD03411999
InChI Key AQOKCDNYWBIDND-FTOWTWDKSA-N
IUPAC Name (5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenylpent-1-en-1-yl]cyclopentyl]-N-ethylhept-5-enamide
SMILES CCNC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)CCC1=CC=CC=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White
Quantity 100 mg
Formula Weight 415.57
Chemical Name or Material Bimatoprost
Hazard Category GHS07: Exclamation mark
Hazard Statement GHS08: Health hazard
ShelfLife 2 years
Recommended Storage Freezer -20°C