missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bimagrumab Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA541992
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Bimagrumab has been used in trials studying the treatment and supportive care of Sarcopenia, Skeletal Muscle, Mechanical Ventilation, Sporadic Inclusion Body Myositis, and Sporadic Inclusion Body Myositis (sIBM), among others.Identifiants chimiques
| Bimagrumab | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG1 λ | |
| Human | |
| BYM338 | |
| Primary | |
| Protein A/G |
| 1.99 mg/mL | |
| ELISA, Flow Cytometry | |
| Unconjugated | |
| Human | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Human ACVR2A/ACVR2B. | |
| Antibody |
Spécifications
| Bimagrumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BYM338 | |
| Human ACVR2A/ACVR2B. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, Flow Cytometry | |
| 1.99 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 λ |