missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ Terminator v1.0 & v3.0 5X Sequencing Buffer

Catalog No. 4305603
Encompass
Change view
Click to view available options
Quantity:
10.8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4305603 10.8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4305603 Supplier Applied Biosystems™ Supplier No. 4305603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.

Optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.

Order Info

10.8mL

Specifications

Buffer Sequencing Buffer
Concentration 5 X
Content And Storage Store at -15 to -25°C
For Use With (Equipment) GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
For Use With (Application) Standard Sequencing
Product Line BigDye Terminator, BigDye
Product Type Sequencing Buffer
Quantity 10.8 mL
Shipping Condition Dry Ice
Form Liquid
Are the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 interchangeable?

The composition of the BigDye Terminator 5X buffer v1.1 & v3.1 and BigDye Terminator 5X buffer v1.0 & v3.0 chemistries is different and therefore should only be used with the matching chemistry. Use of the incorrect BigDye Terminator 5X buffer can negatively impact sequencing results.

For Research Use Only. Not for use in diagnostic procedures.