missing translation for 'onlineSavingsMsg'
Learn More

Bicyclohexyl, 99%

Catalog No. AC402211000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402211000 100 g Glass bottle
AC402210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402211000 Supplier Thermo Scientific Chemicals Supplier No. 402211000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-51-3 | C12H22 | 166.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-51-3
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003815
InChI Key WVIIMZNLDWSIRH-UHFFFAOYSA-N
Synonym bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro
PubChem CID 7094
IUPAC Name cyclohexylcyclohexane
SMILES C1CCC(CC1)C1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 3.0°C to 4.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 227.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4780 to 1.4800
Quantity 100 g
Beilstein 05, 108
Specific Gravity 0.86
Solubility Information Solubility in water: slightly soluble.
Formula Weight 166.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bicyclohexyl, 99%
Show More Show Less
EINECSNumber 202-161-4
TSCA TSCA

RUO – Research Use Only