Learn More
Foxx Life Sciences EZBio Ultra Bag Assembly, Single Use BioProcess, 2D, 2L, PE Bag, 2x 3/8"" port with Plug, 1x1/4""HB with needleless Luer insert, CFlex TPE Tubing, Sterile, 10/CS
Numéro de catalogue. 502886451
Encompass
Foxx Life Sciences offers its high-performance Single-Use Bag Assembly for efficient fluid storage and transfer in critical bioprocessing. Designed for optimized flow with minimal fluid hold-up, these bags ensure reliable, high-throughput performance. Made from high-clarity medical-grade Renolit 9101 PE multilayer film, they meet ISO & USP biocompatibility standards, offering low oxygen/CO permeability and excellent low-temperature performance.
Available in sizes 50mL50L with edge ports and 20L and 50L with face ports, customizable up to 4 edge ports, double-bagged, and gamma-sterilized in the U.S.
Features
Optimized ports for enhanced flow rates & integrity
Customizable MTO assemblies with multiple port configurations
Low-profile design minimizes product hold-up
Applications
Buffer and media storage
Bulk product collection and transport
Chromatography and filtration buffer management
Fraction collection and product sampling
Numéro de catalogue. 502886451 Fournisseur Foxx Life Sciences Code fournisseur 66P511117FLS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Foxx Life Sciences offers its high-performance Single-Use Bag Assembly for efficient fluid storage and transfer in critical bioprocessing. Designed for optimized flow with minimal fluid hold-up, these bags ensure reliable, high-throughput performance. Made from high-clarity medical-grade Renolit 9101 PE multilayer film, they meet ISO & USP biocompatibility standards, offering low oxygen/CO permeability and excellent low-temperature performance.
Available in sizes 50mL50L with edge ports and 20L and 50L with face ports, customizable up to 4 edge ports, double-bagged, and gamma-sterilized in the U.S.
Features
Optimized ports for enhanced flow rates & integrity
Customizable MTO assemblies with multiple port configurations
Low-profile design minimizes product hold-up
Applications
Buffer and media storage
Bulk product collection and transport
Chromatography and filtration buffer management
Fraction collection and product sampling