missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BETP
Description
Selective positive allosteric modulator and partial agonist of the glucagon-like peptide 1 (GLP-1) receptor. Increases binding affinity of oxyntomodulin for the GLP-1 receptor. Potentiates oxyntomodulin-mediated GLP-1 receptor signaling in vitro and insulin secretion in vivo. Has no effect on GLP-2, GIP, PTH or glucagon receptors.
Spécifications
Spécifications
| Quantity | 10 mg |
| Formula Weight | 406.42 |
| Molecular Weight (g/mol) | 406.423 |
| Chemical Name or Material | BETP |
Sécurité et manipulation
| Recommended Storage | Store at +4°C |