missing translation for 'onlineSavingsMsg'
Learn More

Betaxolol Hydrochloride 98.0+%, TCI America™

Catalog No. B447450MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B447450MG 50 mg
B4474250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B447450MG Supplier TCI America Supplier No. B447450MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63659-19-8
Molecular Formula C18H30ClNO3
Molecular Weight (g/mol) 343.892
MDL Number MFCD00242959
InChI Key CHDPSNLJFOQTRK-UHFFFAOYSA-N
Synonym betaxolol hydrochloride, betaxolol hcl, betoptima, kerlone, betoptic, kerlon, lokren, betaxolo hydrochloride, betoptic s
PubChem CID 107952
ChEBI CHEBI:643228
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 50 mg
Formula Weight 343.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Betaxolol Hydrochloride
RTECSNumber UA9823000
TSCA No