missing translation for 'onlineSavingsMsg'
Learn More

Betaine monohydrate, 99+%, for analysis

Catalog No. AC402195000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402195000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402195000 Supplier Thermo Scientific Chemicals Supplier No. 402195000
Only null left

CAS: 590-47-6 | C5H13NO3 | 135.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 590-47-6 , 107-43-7
Molecular Formula C5H13NO3
Molecular Weight (g/mol) 135.16
MDL Number MFCD00150010
InChI Key NJZRLXNBGZBREL-UHFFFAOYSA-N
Synonym betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496
PubChem CID 134128033
IUPAC Name carboxymethyl(trimethyl)azanium;hydroxide;hydrate
SMILES [OH-].C[N+](C)(C)CC(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Loss on Drying 11% to 13.5% (140°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3)3NCH2CO2H·OH
Quantity 500 g
Beilstein 04, 346
Merck Index 15, 1181
Solubility Information (10% in H2O) Clear colorless solution
Transmittance 95% min. (c=10, 430nm) 1cm cell
Formula Weight 135.16
Percent Purity 99+%
Grade Analysis
Physical Form Crystalline Powder or Crystals
Chemical Name or Material βine monohydrate
Show More Show Less
EINECSNumber 209-684-7

RUO – Research Use Only