missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 18 Guard Cartridges

Catalog No. 71505014001
Change view
Click to view available options
Diameter (Metric):
4 mm
Length (Metric):
10 mm
Particle Size:
5 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric) Particle Size
71505014001 4 mm 10 mm 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71505014001 Supplier Thermo Scientific™ Supplier No. 71505014001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Available in limited dimensions. Please contact Customer Service for more details.

Thermo Scientific™ BetaBasic™ 18 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Requires:

Thermo Scientific Unigard™ direct connection or stand-alone holder, sold separately.

Specifications

Carbon Load 13%
pH 2 to 12
Particle Size 5 μm
Length (Metric) 10 mm
Pore Size 150Å
Surface Area 200 m2/g
Diameter (Metric) 4 mm
For Use With (Equipment) Uniguard Direct-connection and Stand Alone Holder
Endcapped Yes
USP Type L1
Product Line Beta
Show More Show Less