missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Nicotinamide Adenine Dinucleotide Phosphate Sodium Salt Hydrate, oxidized form 90.0+%, TCI America™
Specifications
Specifications
| Melting Point | 178°C |
| Color | White-Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 1 g |
| Formula Weight | 766.40 |
| Percent Purity | ≥90.0% (HPLC) |
| Grade | Not Available |
| Chemical Name or Material | beta-Nicotinamide Adenine Dinucleotide Phosphate Sodium Salt Hydrate, oxidized form [for Biochemical |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |