missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide Adenine Dinucleotide Phosphate 83.0+%, TCI America™

Catalog No. N0943100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0943100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0943100MG Supplier TCI America Supplier No. N0943100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53-59-8
Molecular Formula C21H30N7O18P3
Molecular Weight (g/mol) 761.423
MDL Number MFCD00067537
InChI Key ZKJOXOJMGXFSPF-QYZPTAICSA-N
Synonym Coenzyme II, beta-NADP, TPN, beta-Triphosphopyridine Nucleotide
PubChem CID 131675342
IUPAC Name [(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methyl [[(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate;hydrate
SMILES C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)(O)OP(=O)([O-])OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 743.41
Percent Purity ≥83.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Nicotinamide Adenine Dinucleotide Phosphate [for Biochemical Research]
RTECSNumber UU3440000
TSCA Yes
Recommended Storage Freezer (-20 °C)