missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthoflavone, 98+%

Catalog No. AAA1854303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1854303 1 g
AAA1854306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1854303 Supplier Thermo Scientific Chemicals Supplier No. A1854303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6051-87-2 | C19H12O2 | 272.303 g/mol

Inhibitor of aryl hydrocarbon hydroxylase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6051-87-2
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.303
MDL Number MFCD00004986
InChI Key OUGIDAPQYNCXRA-UHFFFAOYSA-N
Synonym beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
PubChem CID 2361
ChEBI CHEBI:77013
IUPAC Name 3-phenylbenzo[f]chromen-1-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43

Specifications

Melting Point 162°C to 166°C
Odor Odorless
Quantity 1 g
Beilstein 18991
Formula Weight 272.31
Percent Purity ≥98%
Chemical Name or Material beta-Naphthoflavone
EINECSNumber 227-958-4
RTECSNumber QL6200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only