missing translation for 'onlineSavingsMsg'
Learn More

beta-Isocupreidine 98,0+%, TCI America™

Catalog No. I07281G
Change view
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
I07281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07281G Supplier TCI America Supplier No. I07281G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 253430-48-7
Formule moléculaire C19H22N2O2
Poids moléculaire (g/mol) 310.397
Clé InChI ULTQUFCPWDEOND-JHQYFNNDSA-N
Synonyme (9S)-3alpha,9-Epoxy-10,11-dihydrocinchonan-6′C-ol, beta-ICD
PubChem CID 91827787
SOURIRES CCC12CN3CCC1CC3C(O2)C4=C5C=C(C=CC5=NC=C4)O

Specifications

Nom ou substance chimique beta-Isocupreidine
Point de fusion 259 °C
Couleur Blanc-jaune
Quantité 1 g
Poids de formule 310.40
Pourcentage de pureté ≥98,0% (HPLC,T)
Forme physique Poudre cristalline