missing translation for 'onlineSavingsMsg'
Learn More

beta-III Tubulin Antibody, R&D Systems™

Catalog No. NB1001612
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001612 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001612 Supplier Bio-Techne Supplier No. NB1001612

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Chicken Polyclonal Antibody has been used in 16 publications

beta-III Tubulin Polyclonal specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, KnockDown, Western Blot.

Specifications

Antigen beta-III Tubulin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), KnockDown, Western Blot
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:10000 - 1:20000, Immunohistochemistry 1:2000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount, KnockDown Validated
Formulation 10 mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide
Gene Accession No. Q13509
Gene Alias beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Gene Symbols TUBB3
Host Species Chicken
Immunogen Chickens were immunized with three synthetic peptide/keyhole limpet hemocyanin (KLH) conjugates. These synthetic peptides corresponded to different regions of the beta-Tubulin 3 gene product, but are shared between the human (NP_AAL28094, NCBI) and rat (AAM28438, NCBI) protein sequences.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381
Test Specificity beta-III Tubulin - Dendrite Marker
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgY
Product Line Novus
Show More Show Less

For Research Use Only